Learn More
Description
Specifications
Specifications
| Antigen | PACSIN2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.02-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Formulation | 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide |
| Gene Alias | cytoplasmic phosphoprotein PACSIN2, protein kinase C and casein kinase substrate in neurons 2, protein kinase C and casein kinase substrate in neurons protein 2, SDPII, syndapin II |
| Gene Symbols | PACSIN2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
