Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PACSIN2 Monoclonal Antibody (1735CT116.76.60)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537727
Description
PACSIN2 Monoclonal Antibody for Western Blot
PACSIN 2, a 486 AA phosphoprotein is a novel member of the PACSIN family of cytoplasmic adapter proteins. It is an isoform of PACSIN with a CDC15 N-terminal domain, a C-terminal SH3 domain, 3 conserved regions specific to the PACSIN family, and 3 asn-pro-phe (NPF) motifs, which potentially bind to EH domains. This protein is known to display a ubiquitous expression pattern with the highest levels of expression in brain. Reports suggest that PACSIN 2 participates in the organization of the actin cytoskeleton, the regulation of vesicular traffic. It also participates in endocytic internalization and endocytic receptor recycling.
Specifications
PACSIN2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UNF0, Q9WVE8 | |
PACSIN2 | |
recombinant protein between 250-486 amino acids from human PACSIN2. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Western Blot | |
1735CT116.76.60 | |
Unconjugated | |
PACSIN2 | |
AI197433; cytoplasmic phosphoprotein PACSIN2; Pacsin2; pacsin2.S; protein kinase C and casein kinase substrate in neurons 2; protein kinase C and casein kinase substrate in neurons 2 protein; protein kinase C and casein kinase substrate in neurons 2 S homeolog; protein kinase C and casein kinase substrate in neurons protein 2; RP3-437M21.1; SdpII; SdpIIsyndapin II; synaptic dynamin-associated protein II; syndapin 2; Syndapin II; syndapin2; syndapin-2; Syndapin-II; XELAEV_18019736mg; x-PACSIN2 | |
Mouse | |
Protein G | |
RUO | |
11252, 23970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction