Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACSIN2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA596804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596804 Supplier Thermo Scientific Supplier No. PA596804

Rabbit Polyclonal Antibody

Immunogen sequence: RLRFFREVLL EVQKHLDLSN VAGYKAIYHD LEQSIRAADA VEDLRWFRAN HGPGMAMNWP QFEEWSADLN RTLSRREKKK ATDGVTLTGI NQTGDQSLPS KPSSTLNVPS NPAQSAQSQS SYNPFEDEDD TGSTVSEKDD TKAKNVSSYE KTQSYPTDWS DDESNNPFSS T; Positive Samples: BT-474, HepG2, Mouse lung, Mouse heart; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle membrane, Early endosome, Membrane, Peripheral membrane protein, Recycling endosome membrane, caveola, cytoskeleton, ruffle membrane

PACSIN 2, a 486 AA phosphoprotein is a novel member of the PACSIN family of cytoplasmic adapter proteins. It is an isoform of PACSIN with a CDC15 N-terminal domain, a C-terminal SH3 domain, 3 conserved regions specific to the PACSIN family, and 3 asn-pro-phe (NPF) motifs, which potentially bind to EH domains. This protein is known to display a ubiquitous expression pattern with the highest levels of expression in brain. Reports suggest that PACSIN 2 participates in the organization of the actin cytoskeleton, the regulation of vesicular traffic. It also participates in endocytic internalization and endocytic receptor recycling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACSIN2
Applications Western Blot
Classification Polyclonal
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene PACSIN2
Gene Accession No. Q9UNF0, Q9WVE8
Gene Alias AI197433, cytoplasmic phosphoprotein PACSIN2, Pacsin2, pacsin2.S, protein kinase C and casein kinase substrate in neurons 2, protein kinase C and casein kinase substrate in neurons 2 protein, protein kinase C and casein kinase substrate in neurons 2 S homeolog, protein kinase C and casein kinase substrate in neurons protein 2, RP3-437M21.1, SdpII, SdpIIsyndapin II, synaptic dynamin-associated protein II, syndapin 2, Syndapin II, syndapin2, syndapin-2, Syndapin-II, XELAEV_18019736mg, x-PACSIN2
Gene Symbols PACSIN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-410 of human PACSIN2 (NP_0091602)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11252, 23970
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.