Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACSIN3 Antibody (OTI4F8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP203475
Description
PACSIN3 Monoclonal specifically detects PACSIN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PACSIN3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
PACSIN3 | |
Full length human recombinant protein of human PACSIN3 (NP_057307) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI4F8 | |
Unconjugated | |
Endophilin I, FLJ17861, protein kinase C and casein kinase substrate in neurons 3, protein kinase C and casein kinase substrate in neurons protein 3, SDPIII, SH3 domain-containing protein 6511, syndapin III | |
Mouse | |
48.3 kDa | |
0.1 mL | |
Cancer, Diabetes Research, GPCR, Neuroscience, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
29763 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction