Learn More
Invitrogen™ PACSIN3 Monoclonal Antibody (OTI4G7)

Description
PACSIN3 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | PACSIN3 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI4G7 |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PACSIN3 |
| Gene Accession No. | Q9UKS6 |
| Gene Alias | 4921507A02Rik; 6330413E15Rik; AW413130; EGK_06300; Endophilin I; PACSIN3; Protein kinase C and casein kinase II substrate protein 3; protein kinase C and casein kinase substrate in neurons 3; protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.