Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACSIN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597139 Supplier Invitrogen™ Supplier No. PA597139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HepG2, MCF-7, HeLa, Jurkat, Mouse heart, Mouse liver, Mouse lung, Rat liver, Rat brain; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein.

This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACSIN3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PACSIN3
Gene Accession No. Q99JB8, Q9UKS6
Gene Alias 4921507A02Rik; 6330413E15Rik; AW413130; EGK_06300; Endophilin I; PACSIN3; Protein kinase C and casein kinase II substrate protein 3; protein kinase C and casein kinase substrate in neurons 3; protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III
Gene Symbols PACSIN3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29763, 311187, 80708
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.