Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529950 Supplier Invitrogen™ Supplier No. PA529950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, Jurkat, NT2D. Predicted reactivity: Mouse (98%), Rat (98%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACT
Applications Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PRKRA
Gene Accession No. O75569
Gene Alias AV120107; DYT16; HSD14; HSD-14; interferon-inducible double stranded RNA-dependent protein kinase activator A; interferon-inducible double-stranded RNA-dependent protein kinase activator A; lear; PACT; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; PRK; PRKRA; protein activator of interferon induced protein kinase EIF2AK2; protein activator of the interferon-induced protein kinase; protein kinase, interferon inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX
Gene Symbols PRKRA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PACT. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8575
Target Species Amphibian, Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.