Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565135 Supplier Invitrogen™ Supplier No. PA565135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLAKHRAAEAA INILKANASI CFAVPDPLMP DPSKQPKNQL NPIGSLQELA IHHGWRLPEY TLSQEGGPAH KREYTTICRL ESFMETGK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRKRA
Gene Accession No. O75569
Gene Alias AV120107; DYT16; HSD14; HSD-14; interferon-inducible double stranded RNA-dependent protein kinase activator A; interferon-inducible double-stranded RNA-dependent protein kinase activator A; lear; PACT; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; PRK; PRKRA; protein activator of interferon induced protein kinase EIF2AK2; protein activator of the interferon-induced protein kinase; protein kinase, interferon inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX
Gene Symbols PRKRA
Host Species Rabbit
Immunogen Recombinant Human PACT. Recombinant protein control fragment (Product #RP-104651).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.