Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACT Rabbit anti-Human, Mouse, Rat, Clone: 2W2E10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | PACT |
---|---|
Clone | 2W2E10 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
PACT Monoclonal antibody specifically detects PACT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PACT | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DYT16interferon-inducible double stranded RNA-dependent activator, PACTPKR-associating protein X, PKR-associated protein X, protein kinase, interferon-inducible double stranded RNA dependent activator, RAXHSD14 | |
A synthetic peptide corresponding to a sequence within amino acids 214-313 of human PACT (O75569). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
2W2E10 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Epigenetics | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8575 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title