Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACT Rabbit anti-Human, Mouse, Rat, Clone: S04-8F7, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | PACT |
---|---|
Clone | S04-8F7 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PACT Monoclonal antibody specifically detects PACT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PACT | |
Western Blot | |
Unconjugated | |
Rabbit | |
Epigenetics | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
8575 | |
IgG | |
Affinity-purified |
S04-8F7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DYT16interferon-inducible double stranded RNA-dependent activator, PACTPKR-associating protein X, PKR-associated protein X, protein kinase, interferon-inducible double stranded RNA dependent activator, RAXHSD14 | |
A synthetic peptide of of human PACT (Uniprot#: O75569) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title