Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PADI1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23776125UL
Description
PADI1 Polyclonal specifically detects PADI1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PADI1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PADI1 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: PSACLKLFQEKKEEGYGEAAQFDGLKHQAKRSINEMLADRHLQRDNLHAQKCIDWN | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.05mg/mL | |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
EC 3.5.3.15, HPAD10, hPAD-colony 10, PAD1, PDI, PDI1Protein-arginine deiminase type I, peptidyl arginine deiminase, type I, Peptidylarginine deiminase I, protein-arginine deiminase type-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
29943 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction