Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PADI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561579 Supplier Invitrogen™ Supplier No. PA561579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWLPKEDCRD EKVYSKEDLK DMSQMILRTK GPDRLPAGYE IVLYISMSDS DKVGVFYVEN PFFGQRYIHI LGRRKLYHVV KYTGGS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type II enzyme is the most widely expressed family member. Known substrates for this enzyme include myelin basic protein in the central nervous system and vimentin in skeletal muscle and macrophages. This enzyme is thought to play a role in the onset and progression of neurodegenerative human disorders, including Alzheimer disease and multiple sclerosis, and it has also been implicated in glaucoma pathogenesis. This gene exists in a cluster with four other paralogous genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PADI2
Gene Accession No. Q9Y2J8
Gene Alias KIAA0994; mKIAA0994; PAD type II; Pad2; PAD-H19; PADI2; Pdi; PDI2; peptidyl arginine deiminase 2; peptidyl arginine deiminase, type 2; peptidyl arginine deiminase, type II; peptidyl arginine deiminase, type II; PAD type II; peptidylarginine deiminase II; Protein-arginine deiminase type II; protein-arginine deiminase type-2; RP1-37C10.6
Gene Symbols PADI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PADI2. Recombinant protein control fragment (Product #RP-103009).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11240
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.