Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PADI3 Monoclonal Antibody (C10), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543630
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543630 100 μL
PIMA543631 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543630 Supplier Invitrogen™ Supplier No. MA543630
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PADI3 Monoclonal antibody specifically detects PADI3 in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI3
Applications Western Blot
Classification Monoclonal
Clone C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PADI3
Gene Accession No. P70708, Q9ULW8
Gene Alias PAD3; PADI3; PDI3; peptidyl arginine deiminase 3; peptidyl arginine deiminase, type III; peptidylarginine deiminase III; Protein-arginine deiminase type III; protein-arginine deiminase type-3
Gene Symbols PADI3
Host Species Mouse
Immunogen Met1-Pro300 (Accession Q9ULW8), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29520, 51702
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.