Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PADI4 Antibody (OTI4H5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273226F
Description
PADI4 Monoclonal specifically detects PADI4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PADI4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PADI4 | |
Full length recombinant protein of human PADI4 (NP_036519) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4H5 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
23569.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction