Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Monoclonal Antibody (OTI5B7), TrueMAB™, OriGene
SDP

Catalog No. 502540047
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0047 30 μL
50-167-5491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0047 Supplier OriGene Supplier No. TA504897S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PADI4 Monoclonal antibody specifically detects PADI4 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PADI4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI5B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Padi4
Gene Accession No. Q9UM07
Gene Alias HL-60 PAD; PAD; PAD type IV; PAD4; Padi4; PADI5; PADI-H protein; PAD-R4; Pdi4; PDI5; peptidyl arginine deiminase 4; peptidyl arginine deiminase, type 4; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; peptidylarginine deiminase IV; Peptidylarginine deiminase type alpha; protein-arginine deiminase type IV; Protein-arginine deiminase type-4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 299-588 of human PADI4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23569
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.