Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Monoclonal Antibody (OTI5E7), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504814

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-5425


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at ′Arg-8′ and/or ′Arg-17′ and histone H4 at ′Arg-3′, which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at ′Arg-2142′, which favors its interaction with NCOA2/GRIP1.
Specifications

Specifications

PADI4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UM07
PADI4
Human recombit protein fragment corresponding to amino acids 299-588 of human PADI4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5E7
Unconjugated
PADI4
PAD, PAD4, PADI5, PDI4, PDI5
Mouse
Affinity Chromatography
RUO
23569
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.