Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PADI4 Monoclonal Antibody (OTI5E7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504814S


Description
PADI4 Monoclonal antibody specifically detects PADI4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PADI4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UM07 | |
Mouse | |
Affinity chromatography | |
RUO | |
23569 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5E7 | |
Unconjugated | |
Padi4 | |
HL-60 PAD; PAD; PAD type IV; PAD4; Padi4; PADI5; PADI-H protein; PAD-R4; Pdi4; PDI5; peptidyl arginine deiminase 4; peptidyl arginine deiminase, type 4; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; peptidylarginine deiminase IV; Peptidylarginine deiminase type alpha; protein-arginine deiminase type IV; Protein-arginine deiminase type-4 | |
Full length human recombinant protein of human PADI4 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction