Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PADI4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534702 Supplier Invitrogen™ Supplier No. PA534702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PAD4-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at 'Arg-8' and/or 'Arg-17' and histone H4 at 'Arg-3', which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at 'Arg-2142', which favors its interaction with NCOA2/GRIP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.01% thimerosal; pH 7
Gene Padi4
Gene Accession No. Q9UM07
Gene Alias HL-60 PAD; PAD; PAD type IV; PAD4; Padi4; PADI5; PADI-H protein; PAD-R4; Pdi4; PDI5; peptidyl arginine deiminase 4; peptidyl arginine deiminase, type 4; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; peptidylarginine deiminase IV; Peptidylarginine deiminase type alpha; protein-arginine deiminase type IV; Protein-arginine deiminase type-4
Gene Symbols Padi4
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PAD4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23569
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.