Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PADI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PADI4 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-100-2667
![]() ![]() |
Proteintech Group Inc
173731AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at ′Arg-8′ and/or ′Arg-17′ and histone H4 at ′Arg-3′, which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at ′Arg-2142′, which favors its interaction with NCOA2/GRIP1.Specifications
PADI4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O88807, Q9UM07, Q9Z183 | |
18602, 23569, 29512 | |
PADI4 Fusion Protein Ag11068 | |
Primary | |
-20°C | |
PADI4 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HL 60 PAD, PAD, PAD4, PADI4, PADI5, PDI4, PDI5, Peptidylarginine deiminase IV | |
Padi4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title