Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980215
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0215 Supplier Bioss Supplier No. BS0318R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at ′Arg-8′ and/or ′Arg-17′ and histone H4 at ′Arg-3′, which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at ′Arg-2142′, which favors its interaction with NCOA2/GRIP1.

Specifications

Antigen PADI4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Padi4
Gene Accession No. Q9UM07
Gene Alias HL-60 PAD; PAD; PAD type IV; PAD4; PADI4; PADI5; PADI-H protein; PAD-R4; Pdi4; PDI5; peptidyl arginine deiminase 4; peptidyl arginine deiminase, type 4; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; Peptidylarginine deiminase IV; Peptidylarginine deiminase type alpha; Protein-arginine deiminase type IV; Protein-arginine deiminase type-4
Gene Symbols Padi4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PADI4 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23569
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.