Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PADI6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572059
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572059 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572059 Supplier Invitrogen™ Supplier No. PA572059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PADI6 Polyclonal Antibody for Western Blot

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. This protein may play a role in cytoskeletal reorganization in the egg and in early embryo development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI6
Applications Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PADI6
Gene Accession No. Q6TGC4, Q8K3V4
Gene Alias arginine deiminase-like protein; Egg and embryo abundant PAD; ePAD; hPADVI; PAD6; Padi5; Padi6; peptidyl arginine deiminase 6; peptidyl arginine deiminase, egg and embryo abundant; peptidyl arginine deiminase, type VI; peptidyl arginine deiminase-like protein; peptidylarginine deiminase type 6; peptidylarginine deiminase VI; Protein-arginine deiminase type VI; protein-arginine deiminase type-6
Gene Symbols PADI6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 196-223 amino acids from the N-terminal region of human PADI6.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242726, 353238
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.