Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAEP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556162 Supplier Invitrogen™ Supplier No. PA556162
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDLELPKLAG TWHSMAMATN NISLMATLKA PLRVHITSLL PTPEDNLEIV LHRWENNSCV EKKVLGEKTE NPKKFKI Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 29%.

Human placental protein 14 (PP14; also known as glycodelin and progesterone-associated endometrial protein) is a protein of the lipocalin structural superfamily. PP14 is the most abundant product of the secretory endometrium, and has been proposed as a biochemical marker of endometrial function in women undergoing in vitro fertilization treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAEP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAEP
Gene Accession No. P09466
Gene Alias alpha uterine protein; GD; GdA; GdF; GdS; Glycodelin; glycodelin-A; glycodelin-F; glycodelin-S; PAEG; PAEP; PEG; PEP; placental protein 14; PP14; PP14 protein (placental protein 14); pregnancy-associated endometrial alpha-2 globulin; pregnancy-associated endometrial alpha-2-globulin; progestagen associated endometrial protein; Progestagen-associated endometrial protein; progestagen-associated endometrial protein (placental protein 14, pregnancy-associated endometrial a; progesterone-associated endometrial protein; ZIF-1; Zona-binding inhibitory factor-1
Gene Symbols PAEP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAEP. Recombinant protein control fragment (Product #RP-92931).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5047
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.