Learn More
Invitrogen™ PAEP Polyclonal Antibody

Description
Immunogen sequence: NYTVANEATL LDTDYDNFLF LCLQDTTTPI QSMMCQYLAR VLVEDDEIMQ GFIRAFRPLP RHLWYLLDLK QME Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
Specifications
Specifications
| Antigen | PAEP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PAEP |
| Gene Accession No. | P09466 |
| Gene Alias | alpha uterine protein; GD; GdA; GdF; GdS; Glycodelin; glycodelin-A; glycodelin-F; glycodelin-S; PAEG; PAEP; PEG; PEP; placental protein 14; PP14; PP14 protein (placental protein 14); pregnancy-associated endometrial alpha-2 globulin; pregnancy-associated endometrial alpha-2-globulin; progestagen associated endometrial protein; Progestagen-associated endometrial protein; progestagen-associated endometrial protein (placental protein 14, pregnancy-associated endometrial a; progesterone-associated endometrial protein; ZIF-1; Zona-binding inhibitory factor-1 |
| Gene Symbols | PAEP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.