Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAF1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600274UV
Description
PAF1 Polyclonal antibody specifically detects PAF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PAF1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
F23149_1, FLJ11123, Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae), Pancreatic differentiation protein 2, PD2hPAF1, RNA polymerase II-associated factor 1 homolog | |
A synthetic peptide which maps to a region between residues 475 and 525 of human RNA Polymerase Associated Factor using the numbering given in TrEMBL entry Q9NUU9 (GeneID 54623). | |
0.1 mL | |
Transcription Factors and Regulators | |
54623 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction