Learn More
PAF1 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 154411AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0230
Only null left
Add to Cart
Description
Description
The MW of this protein is 80 kDa, and this antibody specifically recognizes the 80 kDa protein.
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
PAF1
Polyclonal
Unconjugated
PAF1
F23149_1, hPAF1, PAF1, PD2
Rabbit
Antigen Affinity Chromatography
RUO
54623
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N7H5
PAF1
PAF1 Fusion Protein Ag7212
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.