Learn More
Invitrogen™ PAF49 Monoclonal Antibody (GT7212)

Description
Positive Control: 293T, A549, H1299, HCT116 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PAF49 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT7212 |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Cd3eap |
| Gene Accession No. | O15446 |
| Gene Alias | 1190028F09; 2610103M17Rik; A34.5; antis; Anti-sense to ERCC-1 protein; antisense to ERCC-1 protein; Ase1; ASE-1; CAST; CD3 epsilon associated protein; antisense to Ercc1; CD3E antigen, epsilon polypeptide associated protein; CD3E antigen, epsilon polypeptide-associated protein; CD3e molecule associated protein; CD3e molecule, epsilon associated protein; Cd3eap; CD3E-associated protein; CD3-epsilon-associated protein; DNA-directed RNA polymerase I subunit RPA34; PAF49; RGD1305916; RNA polymerase I-associated factor PAF49 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.