Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAF49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527632 Supplier Invitrogen™ Supplier No. PA527632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, H1299, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Isoform 1 is involved in UBTF-activated transcription, presumably at a step following PIC formation.; Isoform 2 has been described as a component of preformed T-cell receptor (TCR) complex. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAF49
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Cd3eap
Gene Accession No. O15446
Gene Alias 1190028F09; 2610103M17Rik; A34.5; antis; Anti-sense to ERCC-1 protein; antisense to ERCC-1 protein; Ase1; ASE-1; CAST; CD3 epsilon associated protein; antisense to Ercc1; CD3E antigen, epsilon polypeptide associated protein; CD3E antigen, epsilon polypeptide-associated protein; CD3e molecule associated protein; CD3e molecule, epsilon associated protein; Cd3eap; CD3E-associated protein; CD3-epsilon-associated protein; DNA-directed RNA polymerase I subunit RPA34; PAF49; RGD1305916; RNA polymerase I-associated factor PAF49
Gene Symbols Cd3eap
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PAF49. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.