Learn More
PAF49 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301294AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Isoform 1 is involved in UBTF-activated transcription, presumably at a step following PIC formation. Isoform 2 has been described as a component of preformed T-cell receptor (TCR) complex.Specifications
| PAF49 | |
| Polyclonal | |
| Unconjugated | |
| CD3EAP | |
| ASE-1, CAST, PAF49, A34.5, CD3 epsilon associated protein, CD3-epsilon-associated protein, CD3E-associated protein, CD3e antigen, epsilon polypeptide associated protein, DNA-directed RNA polymerase I subunit RPA34, RNA polymerase I-associated factor PAF49, antisense to ERCC 1, antis | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10849 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O15446 | |
| CD3EAP | |
| Between 225 and 275 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.