Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAFAH Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 155261AP150UL

Description
PLA2G7 encodes a protein that is a secreted enzyme that catalyzes the degradation of platelet-activating factor to biologically inactive products. Defects in this gene are a cause of platelet-activating factor acetylhydrolase deficiency.Specifications
PAFAH | |
Polyclonal | |
Unconjugated | |
PLA2G7 | |
Group VIIA phospholipase A2, gVIIA PLA2, LDL PLA(2), LDL PLA2, Lp-PLA2, PAF 2 acylhydrolase, PAF acetylhydrolase, PAFAH, PAF-AH, PLA2G7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27226, 7941 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13093, Q60963 | |
Pla2g7 | |
PAFAH Fusion Protein Ag7227 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction