Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAFAH1B2 Antibody - Azide and BSA Free

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB12015875
Description
PAFAH1B2 Polyclonal specifically detects PAFAH1B2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
PAFAH1B2 | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
PAFAH1B2 | |
Synthetic peptide, corresponding to amino acids 1-229 of PAFAH1B2. | |
0.05 mg | |
Primary | |
PAFAH1B2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1.0 mg/mL | |
Western Blot, ELISA | |
EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
Chicken | |
Affinity Purified | |
RUO | |
5049 | |
Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction