Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAFAH1B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531152 Supplier Invitrogen™ Supplier No. PA531152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, NIH-3T3, JC. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (86%), Chicken (93%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha, beta, and gamma PAFAH subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAFAH1B2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PAFAH1B2
Gene Accession No. P68402, Q61206
Gene Alias AI747451; AU021353; epididymis secretory protein Li 303; HEL-S-303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; mus[b]; PAF acetylhydrolase 30 kDa subunit; PAF-AH 30 kDa subunit; PAF-AH alpha 2; PAFAH subunit beta; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; Pafah1b2; Pafahb; pl; platelet activating factor acetylhydrolase 1b catalytic subunit 2; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase alpha 2 subunit; platelet-activating factor acetylhydrolase alpha 2 subunit (PAF-AH alpha 2); platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1b, alpha2 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, beta subunit; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Symbols PAFAH1B2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PAFAH1B2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18475, 5049
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.