Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAFAH1B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562494
Description
Immunogen sequence: QLINTRQPQA KIIVLGLLPR GEKPNPLRQK NAKVNQLLKV SLPKLANVQL LDTDG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha, beta, and gamma PAFAH subunits.
Specifications
PAFAH1B2 | |
Polyclonal | |
Unconjugated | |
PAFAH1B2 | |
AI747451; AU021353; epididymis secretory protein Li 303; HEL-S-303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; mus[b]; PAF acetylhydrolase 30 kDa subunit; PAF-AH 30 kDa subunit; PAF-AH alpha 2; PAFAH subunit beta; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; Pafah1b2; Pafahb; pl; platelet activating factor acetylhydrolase 1b catalytic subunit 2; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase alpha 2 subunit; platelet-activating factor acetylhydrolase alpha 2 subunit (PAF-AH alpha 2); platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1b, alpha2 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, beta subunit; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5049 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P68402 | |
PAFAH1B2 | |
Recombinant protein corresponding to Human PAFAH1B2. Recombinant protein control fragment (Product #RP-101455). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction