Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAFAH1B3 Antibody (OTI5D2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273229R
Description
PAFAH1B3 Monoclonal specifically detects PAFAH1B3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PAFAH1B3 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PAFAH1B3 | |
Full length recombinant protein of human PAFAH1B3 (NP_001139411) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI5D2 | |
Western Blot | |
EC 3.1.1.47, FLJ44990, PAF acetylhydrolase 29 kDa subunit, PAF-AH 29 kDa subunit, PAFAH subunit gamma, PAF-AH subunit gamma, PAF-AH1b alpha 1 subunit, PAFAHG, platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa), platelet-activating factor acetylhydrolase IB subunit gamma, platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD), platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa) | |
Mouse | |
Affinity Purified | |
Neuroscience | |
5050.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction