Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAFAH1B3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544558
Description
Peptide sequence: GHTAEQVTGG IKAIVQLVNE RQPQARVVVL GLLPRGQHPN PLREKNRQVN Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene encodes an acetylhydrolase that catalyzes the removal of an acetyl group from the glycerol backbone of platelet-activating factor. The encoded enzyme is a subunit of the platelet-activating factor acetylhydrolase isoform 1B complex.
Specifications
| PAFAH1B3 | |
| Polyclonal | |
| Unconjugated | |
| PAFAH1B3 | |
| mus[g]; PAF acetylhydrolase 29 kDa subunit; PAF-AH 29 kDa subunit; PAF-AH alpha 1; PAF-AH gamma; PAFAH subunit gamma; PAF-AH subunit gamma; PAF-AH1b alpha 1 subunit; PAFAH1B3; Pafahg; platelet activating factor acetylhydrolase 1b catalytic subunit 3; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa); platelet-activating factor acetylhydrolase alpha 1 subunit; Platelet-activating factor acetylhydrolase IB subunit gamma; platelet-activating factor acetylhydrolase, isoform 1b, alpha1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 3; platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit; platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD); platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5050 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15102 | |
| PAFAH1B3 | |
| Synthetic peptide directed towards the middle region of human PAFAH1B3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction