Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAFAH1B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588934 Supplier Invitrogen™ Supplier No. PA588934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGEENPASK PTPVQDVQGD GRWMSLHHRF VADSKDKEPE VVFIGDSLVQ LMHQCEIWRE LFSPLHALNF GIGGDGTQHV LWRLENGELE HIRPKIVVVW VGTNNHGHTA EQVTGGIKAI VQLVNERQPQ ARVVVLGLLP RGQHPNPLRE KNRQVNELVR AALAGHPRAH FLDADPGFVH SDGTISHHDM YDYLHLSRLG YTPVCRALHS LLLRLLAQDQ GQGAPLLEPA P; Positive Samples: U-87MG, HeLa, Jurkat; Cellular Location: Cytoplasm.

This gene encodes an acetylhydrolase that catalyzes the removal of an acetyl group from the glycerol backbone of platelet-activating factor. The encoded enzyme is a subunit of the platelet-activating factor acetylhydrolase isoform 1B complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAFAH1B3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PAFAH1B3
Gene Accession No. Q15102
Gene Alias mus[g]; PAF acetylhydrolase 29 kDa subunit; PAF-AH 29 kDa subunit; PAF-AH alpha 1; PAF-AH gamma; PAFAH subunit gamma; PAF-AH subunit gamma; PAF-AH1b alpha 1 subunit; PAFAH1B3; Pafahg; platelet activating factor acetylhydrolase 1b catalytic subunit 3; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa); platelet-activating factor acetylhydrolase alpha 1 subunit; Platelet-activating factor acetylhydrolase IB subunit gamma; platelet-activating factor acetylhydrolase, isoform 1b, alpha1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 3; platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit; platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD); platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Gene Symbols PAFAH1B3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-231 of human PAFAH1B3 (NP_0025641).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5050
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.