Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAFAH1B3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119348
This item is not returnable.
View return policy
Description
PAFAH1B3 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes an acetylhydrolase that catalyzes the removal of an acetyl group from the glycerol backbone of platelet-activating factor. The encoded enzyme is a subunit of the platelet-activating factor acetylhydrolase isoform 1B complex.
Specifications
PAFAH1B3 | |
Polyclonal | |
Unconjugated | |
PAFAH1B3 | |
mus[g]; PAF acetylhydrolase 29 kDa subunit; PAF-AH 29 kDa subunit; PAF-AH alpha 1; PAF-AH gamma; PAFAH subunit gamma; PAF-AH subunit gamma; PAF-AH1b alpha 1 subunit; PAFAH1B3; Pafahg; platelet activating factor acetylhydrolase 1b catalytic subunit 3; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa); platelet-activating factor acetylhydrolase alpha 1 subunit; Platelet-activating factor acetylhydrolase IB subunit gamma; platelet-activating factor acetylhydrolase, isoform 1b, alpha1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 3; platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit; platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD); platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa) | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q15102 | |
PAFAH1B3 | |
Recombinant protein Platelet Activating Factor Acetylhydrolase Ib3. The antigen corresponds to amino acid range 28-231 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction