Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAFAH2 Monoclonal Antibody (1A8)
GREENER_CHOICE

Catalog No. PIMA524418
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524418 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524418 Supplier Invitrogen™ Supplier No. MA524418
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LMKKICAQHE QSRIITVLGS VHRSQTDFAF VTGNLIGKFF STETRGSLDP YEGQEVMVRA MLAFLQKHLD LKEDYNQWNN LIEGIGPSLT PGAPHHLSSL.

Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAFAH2
Applications ELISA
Classification Monoclonal
Clone 1A8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PAFAH2
Gene Accession No. Q99487
Gene Alias 2310074E22Rik; AI507170; HSD-PLA2; PAFAH2; platelet activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2 40kDa; platelet-activating factor acetylhydrolase 2, 40kDa; platelet-activating factor acetylhydrolase 2, cytoplasmic; SD-PLA2; serine-dependent phospholipase A2
Gene Symbols PAFAH2
Host Species Mouse
Immunogen PAFAH2 (NP_000428, 293 a.a. ~ 392 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5051
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.