Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAFAH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100168
Description
Antibody detects endogenous levels of total PAFAH2.
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
Specifications
PAFAH2 | |
Polyclonal | |
Unconjugated | |
PAFAH2 | |
2310074E22Rik; AI507170; HSD-PLA2; PAFAH2; platelet activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2 40kDa; platelet-activating factor acetylhydrolase 2, 40kDa; platelet-activating factor acetylhydrolase 2, cytoplasmic; SD-PLA2; serine-dependent phospholipase A2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100163, 313611, 5051 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P83006, Q8VDG7, Q99487 | |
PAFAH2 | |
A synthesized peptide derived from human PAFAH2(Accession Q99487), corresponding to amino acid residues A245-K295. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction