Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAFAH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550562 Supplier Invitrogen™ Supplier No. PA550562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PAFAH2 protein.

Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAFAH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PAFAH2
Gene Accession No. Q99487
Gene Alias 2310074E22Rik; AI507170; HSD-PLA2; PAFAH2; platelet activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2 40kDa; platelet-activating factor acetylhydrolase 2, 40kDa; platelet-activating factor acetylhydrolase 2, cytoplasmic; SD-PLA2; serine-dependent phospholipase A2
Gene Symbols PAFAH2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human platelet-activating factor acetylhydrolase 2, 40kDa.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5051
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.