Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAG1 Antibody (MEM-255), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500342PEATT594
Description
PAG1 Monoclonal specifically detects PAG1 in Human samples. It is validated for Flow Cytometry.Specifications
PAG1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
PAG1 | |
Recombinant intracellular fragment, corresponding to amino acids 97-432 of Human Cbp/PAG (NM_018440.3). | |
0.1 mL | |
Primary | |
The antibody MEM-255 recognizes an epitope (aa 235-280) of Csk-binding protein (Cbp) located in the cytoplasmic domain, also known as protein associated with glycosphingolipid-enriched microdomains (PAG). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MEM-255 | |
Flow Cytometry | |
CBPMGC138364, Csk-binding protein, PAGFLJ37858, phosphoprotein associated with glycosphingolipid microdomains 1, phosphoprotein associated with glycosphingolipid-enriched microdomains 1, Transmembrane adapter protein PAG, Transmembrane phosphoprotein Cbp | |
Mouse | |
Protein A purified | |
Cancer, Cell Biology, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Transcription Factors and Regulators | |
55824.0 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction