Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAG1 Antibody (PAG-C1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500487H
Description
PAG1 Monoclonal specifically detects PAG1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PAG1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PAG1 | |
C-terminal peptide (last 15 amino acids) of human Csk binding protein coupled to KLH. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Transcription Factors and Regulators | |
55824 | |
Human, Mouse, Rat, Bovine | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PAG-C1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CBPMGC138364, Csk-binding protein, PAGFLJ37858, phosphoprotein associated with glycosphingolipid microdomains 1, phosphoprotein associated with glycosphingolipid-enriched microdomains 1, Transmembrane adapter protein PAG, Transmembrane phosphoprotein Cbp | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody PAG-C1 recognizes an epitope located in the C-terminal domain of Csk-binding protein (Cbp PAG), a 46 kDa ubiquitously expressed transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction