Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAG3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | PAG3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1.0-2.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224614SS
![]() |
Novus Biologicals
NBP224614SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224614
![]() |
Novus Biologicals
NBP224614 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
PAG3 Polyclonal specifically detects PAG3 in Human, Mouse, Rat, Opossum, Primate samples. It is validated for Western Blot.Specifications
PAG3 | |
Western Blot 1.0-2.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Opossum, Primate | |
O43150 | |
8853 | |
A portion of amino acids 700-750 of human ASAP2 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cellular Markers, Cytoskeleton Markers | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2, ArfGAP with SH3 domain, ankyrin repeat and PH domain 2, centaurin, beta 3, CENTB3, DDEF2FLJ42910, development and differentiation enhancing factor 2, Development and differentiation-enhancing factor 2, KIAA0400AMAP2, Pap-alpha, PAPPAG3, Paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, Pyk2 C-terminus-associated protein, SHAG1 | |
ASAP2 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_c and CRA_e), chimpanzee and monkey, 86% homologous in horse, 83% homologous in mouse, rat, and cow, and 73% homologous in dog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title