Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAGE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551940 Supplier Invitrogen™ Supplier No. PA551940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EESSDEQPDE VESPTQSQDS TPAEEREDEG ASAAQGQEPE ADSQELVQPK TGCELGDGPD TKRVCLRNEE QMKLPAEGPE PEADSQEQVH PKTGCERGDG PDVQELGLPN PEEVKTPEED E Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 29%.

PAGE1 belongs to a family that are expressed in a variety of tumors but not in normal tissues, except for the testis. Nothing is presently known about the function of this protein.This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. Unlike the other gene family members, this gene does not encode an antigenic peptide. Nothing is presently known about the function of this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAGE1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAGE1
Gene Accession No. O75459
Gene Alias AL5; CT16.3; G antigen 9; G antigen family B member 1; G antigen, family B, 1 (prostate associated); GAGE9; GAGE-9; GAGEB1; P antigen family member 1; P antigen family, member 1 (prostate associated); PAGE family member 1; PAGE1; PAGE-1; prostate associated gene 1; prostate-associated gene 1 protein
Gene Symbols PAGE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAGE1. Recombinant protein control fragment (Product #RP-92166).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.