Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAGE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550471
Description
The antibody detects endogenous levels of total PAGE2 protein.
PAGE-2 (prostate-associated gene 2 protein), also known as P antigen family member 2B, GAGEC2, CT16.4 or GAGEE2, is a 111 amino acid protein that is expressed primarily in testis and belongs to the GAGE family. PAGE-2B (prostate-associated gene 2B protein), also designated putative G antigen family E member 3, P antigen family member 2B, CT16.5 or GAGEE3, also consists of 111 amino acids and is a member of the GAGE family.
Specifications
PAGE2 | |
Polyclonal | |
Unconjugated | |
PAGE2 | |
CT16.4; G antigen family C 2; g antigen family E member 2; G antigen, family C, 2; G antigen, family E, 2; GAGEC2; GAGEE2; P antigen family member 2; P antigen family, member 2 (prostate associated); PAGE family member 2; PAGE2; PAGE-2; Prostate-associated gene 2 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
203569 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7Z2X7 | |
PAGE2 | |
Full length fusion protein of Human PAGE2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction