Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAGE4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542146 Supplier Invitrogen™ Supplier No. PA542146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DGQEAPDVVA FVAPGESQQE EPPTDNQDIE PGQEREGTPP IEERKVEGDC Sequence homology: Human: 100%.

This gene is a member of the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in prostate and prostate cancer. It is also expressed in other male and female reproductive tissues including testis, fallopian tube, uterus, and placenta, as well as in testicular cancer and uterine cancer. The protein encoded by this gene shares sequence similarity with other GAGE/PAGE proteins, and also belongs to a family of CT (cancer-testis) antigens. The protein may play a role in benign and malignant prostate diseases. A related pseudogene is located on chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAGE4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PAGE4
Gene Accession No. O60829
Gene Alias CT16.7; FLJ35184; G antigen family C member 1; GAGE-9; GAGEC1; JM27; JM-27; P antigen family member 4; P antigen family, member 4 (prostate associated); PAGE family member 4; PAGE-1; PAGE4; PAGE-4; prostate-associated gene protein 4
Gene Symbols PAGE4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PAGE4 (aa 14-63).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9506
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.