Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAGE4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542147
Description
Peptide sequence: PPIEERKVEG DCQEMDLEKT RSERGDGSDV KEKTPPNPKH AKTKEAGDGQ Sequence homology: Human: 100%.
This gene is a member of the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in prostate and prostate cancer. It is also expressed in other male and female reproductive tissues including testis, fallopian tube, uterus, and placenta, as well as in testicular cancer and uterine cancer. The protein encoded by this gene shares sequence similarity with other GAGE/PAGE proteins, and also belongs to a family of CT (cancer-testis) antigens. The protein may play a role in benign and malignant prostate diseases. A related pseudogene is located on chromosome 7.
Specifications
| PAGE4 | |
| Polyclonal | |
| Unconjugated | |
| PAGE4 | |
| CT16.7; FLJ35184; G antigen family C member 1; GAGE-9; GAGEC1; JM27; JM-27; P antigen family member 4; P antigen family, member 4 (prostate associated); PAGE family member 4; PAGE-1; PAGE4; PAGE-4; prostate-associated gene protein 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9506 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60829 | |
| PAGE4 | |
| Synthetic peptide directed towards the middle region of human PAGE4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction