Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PageRuler™ Prestained Protein Ladder, 10 to 180 kDa
GSA_VA

Catalog No. pi26617
Click to view available options
Quantity:
2 x 250 μL
10 x 250 μL

Catalog No. PI26617

Supplier: Thermo Scientific™ 26617

Only null left
Add to Cart
Add to Cart

Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting.

Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Contents: ten vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

Detection Method Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Number of Markers 10
Product Type Protein Ladder
Ready to Load Yes
Size Range 10 to 180 kDa
Stain Type 3 colors: Blue, Orange, Green
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 180, 130, 100, 70, 55, 40, 35, 25, 15, 10 kDa
Product Line PageRuler
Quantity 10 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
System Type Western Blotting, SDS-PAGE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.