Learn More
Invitrogen™ PAGR1 Polyclonal Antibody

Description
A suggested positive control is EL4 cell lysate. PA5-20828 can be used with blocking peptide PEP-0942.
Specifications
Specifications
| Antigen | PAGR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PAGR1 |
| Gene Accession No. | Q99L02, Q9BTK6 |
| Gene Alias | 2900092E17Rik; C16orf53; C77040; GAS; glutamate-rich coactivator associated with SRC1; glutamate-rich coactivator interacting with SRC1; glutamate-rich coactivator interacting with SRC1/NCOA1; PA1; Pagr; PAGR1; Pagr1a; PAXIP1 associated glutamate rich protein 1; PAXIP1 associated glutamate rich protein 1A; PAXIP1 associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1A; PAXIP1-associated protein 1; PTIP-associated 1; PTIP-associated 1 protein; PTIP-associated protein 1 |
| Gene Symbols | PAGR1, Pagr1a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.