Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAH Mixture (For EPA 625, 8270C/Appendix IX)

Catalog No. usus106n
Encompass
Click to view available options
Quantity:
1 x 1 mL

Specifications

Chemical Name or Material PAH Mixture
Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methylene Chloride/Benzene (1:1):
Acenaphthene
Acenaphthylene
Anthracene
Benz[a]anthracene
Benzo[b]fluoranthene
Benzo[k]fluoranthene
Benzo[ghi]perylene
Benzo[a]pyrene
Chrysene
Dibenz[a,h]anthracene
Fluoranthene
Fluorene
Indeno[1,2,3-cd]pyrene
Naphthalene
Phenanthrene
Pyrene
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.