Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAH Mouse anti-Human, Mouse, Rat, Clone: 717CT21.1.1, Abnova™

Catalog No. 89934721 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-721 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-721 Supplier Abnova Corporation Supplier No. MAB12350
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human PAH.

PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria. [provided by RefSeq]

Specifications

Antigen PAH
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 717CT21.1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human PAH.
Dilution Immunohistochemistry (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias PH/PKU/PKU1
Gene Symbols PAH
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 127-161 at internal region of human PAH.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5053
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.